EMAIL THIS PAGE TO A FRIEND
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.]
Commentary: Serum lidocaine levels and cutaneous side effects after application of 23% lidocaine/7% tetracaine ointment to the face.
PMID 23301785