EMAIL THIS PAGE TO A FRIEND
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.]
Letter: Immunocryotherapy for difficult-to-treat basal cell carcinoma of the eyelid.
PMID 23301789