EMAIL THIS PAGE TO A FRIEND
Preparative biochemistry & biotechnology
One-pot synthesis of 2,4,5-trisubstituted imidazoles using [BPy]H2PO4, an efficient and recyclable catalyst.
PMID 23302106
Abstract
[BPy]H(2)PO(4) was easily prepared and used as an efficient and recyclable catalyst for the one-pot synthesis of 2,4,5-trisubstituted imidazoles under solvent-free conditions in good to excellent yields. Solvent-free conditions, simple experimental and workup procedures, and the use of a nontoxic, recyclable catalyst of an environmentally benign nature are remarkable features of the procedure.