EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Design, synthesis and biological evaluation of novel 7-mercaptocoumarin derivatives as α(1)-adrenoceptor antagonists.
PMID 23302583
Abstract
Study on the pharmacophore model of α(1)-adrenoceptor (α(1)-AR) antagonists led to design a series of novel 7-mercaptocoumarin derivatives as α(1)-AR antagonists. All designed compounds have been synthesized and biologically evaluated. The results showed that most of them exhibited strong antagonistic activity. Especially compound 6 showed excellent activity, which was better than that of the reference compound prazosin. Structure-activity relationship studies revealed that small hydrophobic group at the terminal heterocyclic ring and ortho substituents on the phenyl ring of phenylpiperazine moiety were the essential structural factors for α(1)-AR antagonistic activity. The pharmacophore modeling studies further clarified their structural contributions to antagonistic activity and also demonstrated that 7-mercaptocoumarin moiety could be a useful scaffold for design of α(1)-AR antagonists.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P30004
1-Phenylpiperazine, 99%
C10H14N2