EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
DNA duplex stabilization in crowded polyanion solutions.
PMID 23302897
Abstract
The melting temperature of duplex DNA is much higher in polyanions than in non-ionic polymers with similar ionic strength, suggesting an additional electrostatic contribution on top of the excluded volume effect.