EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Highly efficient and chemoselective α-iodination of acrylate esters through Morita-Baylis-Hillman-type chemistry.
PMID 23303274
Abstract
The chemoselective α-iodination of various simple and multi-functionalised acrylic esters is efficiently accomplished by a Morita-Baylis-Hillman protocol involving the use of N-iodophthalimide, 3-quinuclidinol and KF-Celite in acetonitrile. No degradation of the obtained compounds was observed under the optimized conditions thus, furnishing α-iodoacrylates suitable for organometallic reactions (i.e. Nozaki-Kishi-Hiyama type coupling).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
253340
3-Quinuclidinol, 99%
C7H13NO