EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Promiscuous catalysis of asymmetric Michael-type additions of linear aldehydes to β-nitrostyrene by the proline-based enzyme 4-oxalocrotonate tautomerase.
PMID 23303727
Abstract
Exploiting catalytic promiscuity: The proline-based enzyme 4-oxalocrotonate tautomerase (4-OT) promiscuously catalyzes asymmetric Michael-type additions of linear aldehydes--ranging from acetaldehyde to octanal--to trans-β-nitrostyrene in aqueous solvent. The presence of 1.4 mol% of 4-OT effected formation of the anticipated γ-nitroaldehydes in fair to good yields with dr values of up to 93:7 and ee values of up to 81 %.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N26806
trans-β-Nitrostyrene, 99%
C8H7NO2