EMAIL THIS PAGE TO A FRIEND
Journal of mass spectrometry : JMS
Multiplex mass spectrometry imaging for latent fingerprints.
PMID 23303752
Abstract
We have previously developed in-parallel data acquisition of orbitrap mass spectrometry (MS) and ion trap MS and/or MS/MS scans for matrix-assisted laser desorption/ionization MS imaging (MSI) to obtain rich chemical information in less data acquisition time. In the present study, we demonstrate a novel application of this multiplex MSI methodology for latent fingerprints. In a single imaging experiment, we could obtain chemical images of various endogenous and exogenous compounds, along with simultaneous MS/MS images of a few selected compounds. This work confirms the usefulness of multiplex MSI to explore chemical markers when the sample specimen is very limited.
Related Materials