EMAIL THIS PAGE TO A FRIEND
Journal of mass spectrometry : JMS
An effective and rapid determination by MALDI/TOF/TOF of methionine sulphoxide content of ApoA-I in type 2 diabetic patients.
PMID 23303753
Abstract
Increased oxidation of low density lipoprotein (LDL) is characteristic of atherosclerosis. In this frame, high density lipoproteins (HDL) play an important role, being able to remove lipid peroxides (LPOs) and cholesterol from oxidized LDL, so exhibiting a protective role against atherosclerosis. A wide range of reactive compounds lead to the oxidation of methionine (Met) residues with the formation of methionine sulphoxide (MetO) in apolipoprotein A-I (ApoA-I). Consequently, the determination of MetO level can give both an evaluation of oxidative stress and the reduced capability of ApoA-I in LPOs and cholesterol transport. For these reasons, the development of analytical methods able to determine the MetO level is surely of interest, and we report here the results obtained by MALDI mass spectrometry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
64430
DL-Methionine sulfoxide, ≥99.0% (NT)
C5H11NO3S
M1126
L-Methionine sulfoxide
C5H11NO3S