EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Gene-specific factors determine mitotic expression and bookmarking via alternate regulatory elements.
PMID 23303784
Abstract
Transcriptional silencing during mitosis is caused by inactivation of critical transcriptional regulators and/or chromatin condensation. Inheritance of gene expression patterns through cell division involves various bookmarking mechanisms. In this report, we have examined the mitotic and post-mitotic expression of the DRA major histocompatibility class II (MHCII) gene in different cell types. During mitosis the constitutively MHCII-expressing B lymphoblastoid cells showed sustained occupancy of the proximal promoter by the cognate enhanceosome and general transcription factors. In contrast, although mitotic epithelial cells were depleted of these proteins irrespectively of their MHCII transcriptional activity, a distal enhancer selectively recruited the PP2A phosphatase via NFY and maintained chromatin accessibility. Based on our data, we propose a novel chromatin anti-condensation role for this element in mitotic bookmarking and timing of post-mitotic transcriptional reactivation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-599
Anti-acetyl-Histone H3 Antibody, Detect acetyl-Histone H3 with Anti-acetyl-Histone H3 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB, ICC, ChIP, ChIP-seq.
06-866
Anti-acetyl-Histone H4 Antibody, Use Anti-acetyl-Histone H4 Antibody (Rabbit Polyclonal Antibody) has been published and validated in ChIP, WB, ICC to detect acetyl-Histone H4 also known as H4 histone family member A, histone 1-H4a.
07-030
Anti-dimethyl-Histone H3 (Lys4) Antibody, Use Anti-dimethyl-Histone H3 (Lys4) Antibody (rabbit polyclonal antibody) is published and validated in ChIP, DB, ICC, IF, WB, PIA, ChIP-seq. to detect dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4).
06-570
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker is a Rabbit Polyclonal Antibody for detection of Histone H3 phosphorylated at serine 10.This highly published Ab, also known as Anti-H3S10p, has been validated in ICC, IP & WB.
05-421
Anti-PP2A Antibody, C subunit, clone 1D6, Detect PP2A using this Anti-PP2A Antibody, C subunit, clone 1D6 validated for use in IC, IP & WB.
07-473
Anti-trimethyl-Histone H3 (Lys4) Antibody, Anti-trimethyl-Histone H3 (Lys4) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated at lysine 4. Also known as Anti-H3K4me3, this highly specific and well published antibody has been validated in ChIP, DB, WB, PIA, ChIP-seq.