EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Deficits in ventromedial prefrontal cortex group 1 metabotropic glutamate receptor function mediate resistance to extinction during protracted withdrawal from an extensive history of cocaine self-administration.
PMID 23303930
Abstract
Anomalies in prefrontal cortex (PFC) function are posited to underpin difficulties in learning to suppress drug-seeking behavior during abstinence. Because group 1 metabotropic glutamate receptors (mGluRs) regulate drug-related learning, we assayed the consequences of extended access to intravenous cocaine (6 h/d; 0.25 mg/infusion for 10 d) on the PFC expression of group 1 mGluRs and the relevance of observed changes for cocaine seeking. After protracted withdrawal, cocaine-experienced animals exhibited a time-dependent intensification of cue-induced cocaine-seeking behavior and an impaired extinction of this behavior. These behavioral phenomena were associated with a time-dependent reduction in mGluR1/5 expression within ventromedial PFC (vmPFC) of cocaine-experienced animals exposed to extinction testing but not in untested ones. Interestingly, pharmacological manipulations of vmPFC mGluR1/5 produced no immediate effects on cue-induced cocaine-seeking behavior but produced residual effects on a subsequent test for cocaine seeking. At 3 d withdrawal, cocaine-experienced rats infused intra-vmPFC with mGluR1/5 antagonists, either before or after an initial test for cocaine seeking, persisted in their cocaine seeking akin to cocaine-experienced rats in protracted withdrawal. Conversely, cocaine-experienced rats infused with an mGluR1/5 agonist before the initial test for cocaine-seeking at 30 d withdrawal exhibited a facilitation of extinction learning. These data indicate that cue-elicited deficits in vmPFC group 1 mGluR function mediate resistance to extinction during protracted withdrawal from a history of extensive cocaine self-administration and pose pharmacological stimulation of these receptors as a potential approach to facilitate learned suppression of drug-seeking behavior that may aid drug abstinence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9753
DL-3,4-Dihydroxyphenyl glycol
C8H10O4