EMAIL THIS PAGE TO A FRIEND
Mediators of inflammation
Increased expression of visfatin in monocytes and macrophages in male acute myocardial infarction patients.
PMID 23304061
Abstract
We demonstrated that visfatin expressed in monocytes and neutrophils and increased their reactivity in male acute ST-segment elevation myocardial infarction patients. Furthermore, visfatin was strongly appeared in lipid rich coronary rupture plaques and macrophages. These results suggest another explanation about leukocytes mediated visfatin that may play a pathogenesis role in coronary vulnerable plaques rupture.