EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of phenanthridine derivatives by microwave-mediated cyclization of o-furyl(allylamino)arenes.
PMID 23305184
Abstract
A novel and efficient synthesis of phenanthridines and aza analogues is reported. The key step is a microwave-mediated intramolecular Diels-Alder cyclization of o-furyl(allylamino)arenes. In the presence of a catalytic amount of acid, the DA-adduct reacts further to give the dihydrophenanthridines, which easily can be oxidized to fully aromatic compounds by air in the presence of UV light or by DDQ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
528919
2-Bromo-4,6-dichloroaniline, 98%
C6H4BrCl2N