EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Fluorescent tris-imidazolium sensors for picric acid explosive.
PMID 23305246
Abstract
Two new anthracene-functionalized fluorescent tris-imidazolium salts have been synthesized, characterized, and proven to be selective sensors for picric acid, which is a common constituent of many powerful explosives. Theoretical studies revealed an unusual ground-state electron transfer from picrate anion to the sensor molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
197378
Picric acid, moistened with water, ≥98%
C6H3N3O7