EMAIL THIS PAGE TO A FRIEND
Journal of physics. Condensed matter : an Institute of Physics journal
Magnetic and micro-Raman studies of hexagonal-DyMnO3.
PMID 23306085
Abstract
Dc-susceptibility measurements and Raman active phonon frequencies of hexagonal DyMnO(3) retrace the Mn(3+) ions antiferromagnetic transition at T(N) ~ 70 K and their spin reorientation at T(SR) ~ 48 K. The temperature evolution of Raman active mode frequencies and their over-hardening are associated with Dy(3+) and Mn(3+) ion displacements below T(N) and with a spin-phonon coupling that involves apical oxygen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
377473
Manganese(II,III) oxide, 97%
Mn3O4