EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Liquid hot water pretreatment of sugarcane bagasse and its comparison with chemical pretreatment methods for the sugar recovery and structural changes.
PMID 23306094
Abstract
Liquid hot water (LHW), dilute hydrochloric acid (HCl) and dilute sodium hydroxide (NaOH) were applied to sugarcane bagasse (SB). Application of the same analytical methods and material balance approaches facilitated meaningful comparisons of glucose and xylose yields from combined pretreatment and enzymatic hydrolysis. All pretreatments enhanced sugar recovery from pretreatment and subsequent enzymatic hydrolysis substantially compared to untreated sugarcane bagasse. Adding Tween80 in the enzymatic hydrolysis process increased the conversion level of glucan/xylan by 0.3-fold, especially for the low pH pretreatment where more lignin was left in the solids. The total sugar recovery from sugarcane bagasse with the coupled operations of pretreatment and 72 h enzymatic digestion reached 71.6% for LHW process, 76.6% for HCl pretreatment and 77.3% for NaOH pretreatment. Different structural changes at the plant tissue, cellular, and cell wall levels might be responsible for the different enzymatic digestibility. Furthermore, a combined LHW and aqueous ammonia pretreatment was proposed to reduce energy input and enhance the sugar recovery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
223913 Ascarite®, Sodium hydroxide-coated silica, 8-20 mesh
223921 Ascarite®, Sodium hydroxide-coated silica, 20-30 mesh
S8045
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
HNaO
306576
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
HNaO
S5881
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
HNaO
221465
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
HNaO
367176
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
HNaO
484024
Sodium hydroxide, reagent grade, 97%, flakes
HNaO
71690
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
HNaO
30620
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
HNaO
06203
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
HNaO
06306
Sodium hydroxide, ≥98.5%, microprills
HNaO
06208
Sodium hydroxide, for the manufacture of mirrors, pellets, Cl <0.001%
HNaO
655104
Sodium hydroxide, reagent grade, 97%, powder
HNaO
901915
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
HNaO
415413
Sodium hydroxide solution, 50% in H2O
HNaO
05211
Sodium hydroxide solution, purum, ≥32%
HNaO
30530
Sodium hydroxide solution, ≥20%
HNaO
S8263
Sodium hydroxide solution, 5.0 M
HNaO