EMAIL THIS PAGE TO A FRIEND
Journal of the National Comprehensive Cancer Network : JNCCN
Multiple myeloma, version 1.2013.
PMID 23307977
Abstract
These NCCN Guidelines Insights highlight the important updates/changes specific to the management of relapsed or progressive disease in the 2013 version of the NCCN Clinical Practice Guidelines in Oncology for Multiple Myeloma. These changes include the addition of new regimens as options for salvage therapy and strategies to mitigate the adverse effects and risks associated with newer regimens for the treatment of multiple myeloma.