EMAIL THIS PAGE TO A FRIEND
Journal of the American Geriatrics Society
Higher serum vitamin D concentration is associated with better balance in older adults with supra-optimal vitamin D status.
PMID 23311560