EMAIL THIS PAGE TO A FRIEND
Organic letters
Direct access to enantioenriched spiroacetals through asymmetric relay catalytic three-component reaction.
PMID 23312017
Abstract
The gold(I)/chiral Brønsted acid relay catalysis enabled a highly stereoselective three-component reaction of salicylaldehydes, anilines, and alkynols to give aromatic spiroacetals in high yields and stereoselectivities.