EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
A modular, energy-based approach to the development of nickel containing molecular electrocatalysts for hydrogen production and oxidation.
PMID 23313415
Abstract
This review discusses the development of molecular electrocatalysts for H2 production and oxidation based on nickel. A modular approach is used in which the structure of the catalyst is divided into first, second, and outer coordination spheres. The first coordination sphere consists of the ligands bound directly to the metal center, and this coordination sphere can be used to control such factors as the presence or absence of vacant coordination sites, redox potentials, hydride donor abilities and other important thermodynamic parameters. The second coordination sphere includes functional groups such as pendent acids or bases that can interact with bound substrates such as H2 molecules and hydride ligands, but that do not form strong bonds with the metal center. These functional groups can play diverse roles such as assisting the heterolytic cleavage of H2, controlling intra- and intermolecular proton transfer reactions, and providing a physical pathway for coupling proton and electron transfer reactions. By controlling both the hydride donor ability of the catalysts using the first coordination sphere and the proton donor abilities of the functional groups in the second coordination sphere, catalysts can be designed that are biased toward H2 production, oxidation, or bidirectional (catalyzing both H2 oxidation and production). The outer coordination sphere is defined as that portion of the catalytic system that is beyond the second coordination sphere. This coordination sphere can assist in the delivery of protons and electrons to and from the catalytically active site, thereby adding another important avenue for controlling catalytic activity. Many features of these simple catalytic systems are good models for enzymes, and these simple systems provide insights into enzyme function and reactivity that may be difficult to probe in enzymes. This article is part of a Special Issue entitled: Metals in Bioenergetics and Biomimetics Systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295396
Hydrogen, ≥99.99%
H2
268259
Nickel, foil, thickness 0.125 mm, ≥99.9%
Ni
266981
Nickel, powder, <50 μm, 99.7% trace metals basis
Ni
268283
Nickel, powder, <1 μm, 99.8% trace metals basis
Ni
577995
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Ni
267074
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Ni
267058
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Ni
357626
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Ni
357634
Nickel, wire, diam. 0.25 mm, ≥99.9%
Ni
767484
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Ni
GF82156912
Nickel, rod, 100mm, diameter 12.0mm, 99%
Ni
GF11598194
Nickel, rod, 100mm, diameter 12.7mm, 99.98%
Ni
GF29576010
Nickel, rod, 100mm, diameter 16mm, 99.99+%
Ni
GF83584143
Nickel, rod, 100mm, diameter 2.0mm, 99.99+%
Ni
GF45193697
Nickel, rod, 100mm, diameter 2.0mm, annealed, 99%
Ni
GF11517948
Nickel, rod, 100mm, diameter 20.0mm, 99%
Ni
GF39205931
Nickel, rod, 100mm, diameter 25.4mm, 99.98%
Ni
GF69167920
Nickel, rod, 100mm, diameter 3.0mm, 99%
Ni
GF70332870
Nickel, rod, 100mm, diameter 3.3mm, 99.99+%
Ni
GF99799968
Nickel, rod, 100mm, diameter 30mm, 99%
Ni
GF90754241
Nickel, rod, 100mm, diameter 40mm, 99%
Ni
GF20561385
Nickel, rod, 100mm, diameter 5.0mm, 99.99+%
Ni
GF90932823
Nickel, rod, 100mm, diameter 6.35mm, 99%
Ni
GF50593002
Nickel, rod, 100mm, diameter 9.5mm, 99%
Ni
GF68355085
Nickel, rod, 200mm, diameter 1.5mm, 99.99+%
Ni
GF66250256
Nickel, rod, 200mm, diameter 10.0mm, 99.99+%
Ni
GF69613825
Nickel, rod, 200mm, diameter 12.0mm, 99%
Ni
GF49774034
Nickel, rod, 200mm, diameter 12.7mm, 99.98%
Ni
GF97332554
Nickel, rod, 200mm, diameter 16mm, 99.99+%
Ni
GF58660746
Nickel, rod, 200mm, diameter 2.0mm, 99.99+%
Ni