EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Determination of phenylephrine in human plasma using ultra-performance liquid chromatography-tandem mass spectrometry.
PMID 23314401
Abstract
This paper described a sensitive and rapid method based on ultra-performance liquid chromatography coupled to tandem mass spectrometry (UPLC-MS/MS) for the determination of phenylephrine in human plasma. Plasma samples were pre-purified by solid-phase extraction (SPE). The chromatographic separation was achieved with BEH HILIC column using a mixture of 10mM pH 3.5 ammonium formate and acetonitrile (10:90, v/v) under isocratic conditions at a flow rate of 0.4 mL/min. The mass spectrometry was carried out using positive electrospray ionization (ESI) and data acquisition was carried out in the multiple reaction monitoring (MRM) mode. The method was fully validated over the concentration range of 10.0-5000 pg/mL. The lower limit of quantification (LLOQ) was 10.0 pg/mL. Inter- and intra-batch precision was less than 15% and the accuracy was within 85-115%. Extraction recovery was 78.5%. Selectivity, matrix effects and stability were also validated. The method was applied to the pharmacokinetic study of phenylephrine hydrochloride in Chinese subjects.