EMAIL THIS PAGE TO A FRIEND
Folia microbiologica
Real encoded genetic algorithm and response surface methodology to optimize production of an indolizidine alkaloid, swainsonine, from Metarhizium anisopliae.
PMID 23315485
Abstract
Response surface methodology (RSM) and artificial neural network-real encoded genetic algorithm (ANN-REGA) were employed to develop a process for fermentative swainsonine production from Metarhizium anisopliae (ARSEF 1724). The effect of finally screened process variables viz. inoculum size, oatmeal extract, glucose, and CaCl2 were investigated through central composite design and were further utilized for training sets in ANN with training and test R values of 0.99 and 0.94, respectively. ANN-REGA was finally employed to simulate the predictive swainsonine production with best evolved media composition. ANN-REGA predicted a more precise fermentation model with 103 % (shake flask) increase in alkaloid production compared to 75.62 % (shake flask) obtained with RSM model upon validation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9263
Swainsonine, synthetic
C8H15NO3
S8195
Swainsonine, from Metarrhizium anisopliae, ≥98% (TLC)
C8H15NO3