EMAIL THIS PAGE TO A FRIEND
Tetrahedron
Synthesis studies on the Melodinus alkaloid meloscine.
PMID 23316092
Abstract
The pentacyclic Melodinus alkaloid (±)-meloscine was synthesized in 19 chemical steps from 2-bromobenzaldehyde through a route featuring an allenyl azide cyclization cascade to deliver the core azabicyclo[3.3.0]octane substructure. Peripheral functionalization of this core included a Tollens-type aldol condensation to set the quaternary center at C(20) and a diastereoselective ring closing metathesis to forge the tetrahydropyridine ring.