EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Inhibition of IspH, a [4Fe-4S]2+ enzyme involved in the biosynthesis of isoprenoids via the methylerythritol phosphate pathway.
PMID 23316732
Abstract
The MEP pathway, which is absent in animals but present in most pathogenic bacteria, in the parasite responsible for malaria and in plant plastids, is a target for the development of antimicrobial drugs. IspH, an oxygen-sensitive [4Fe-4S] enzyme, catalyzes the last step of this pathway and converts (E)-4-hydroxy-3-methylbut-2-en-1-yl diphosphate (HMBPP) into the two isoprenoid precursors: isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP). A crucial step in the mechanism of this enzyme is the binding of the C4 hydroxyl of HMBPP to the unique fourth iron site in the [4Fe-4S](2+) moiety. Here, we report the synthesis and the kinetic investigations of two new extremely potent inhibitors of E. coli IspH where the OH group of HMBPP is replaced by an amino and a thiol group. (E)-4-Mercapto-3-methylbut-2-en-1-yl diphosphate is a reversible tight-binding inhibitor of IspH with K(i) = 20 ± 2 nM. A detailed kinetic analysis revealed that (E)-4-amino-3-methylbut-2-en-1-yl diphosphate is a reversible slow-binding inhibitor of IspH with K(i) = 54 ± 19 nM. The slow binding behavior of this inhibitor is best described by a one-step mechanism with the slow step consisting of the formation of the enzyme-inhibitor (EI) complex.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7500
meso-Erythritol, ≥99%
C4H10O4