EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Simplifying nickel(0) catalysis: an air-stable nickel precatalyst for the internally selective benzylation of terminal alkenes.
PMID 23316879
Abstract
The synthesis and characterization of the air-stable nickel(II) complex trans-(PCy(2)Ph)(2)Ni(o-tolyl)Cl is described in conjunction with an investigation of its use for the Mizoroki-Heck-type, room temperature, internally selective coupling of substituted benzyl chlorides with terminal alkenes. This reaction, which employs a terminal alkene as an alkenylmetal equivalent, provides rapid, convergent access to substituted allylbenzene derivatives in high yield and with regioselectivity greater than 95:5 in nearly all cases. The reaction is operationally simple, can be carried out on the benchtop with no purification or degassing of solvents or reagents, and requires no exclusion of air or water during setup. Synthesis of the precatalyst is accomplished through a straightforward procedure that employs inexpensive, commercially available reagents, requires no purification steps, and proceeds in high yield.
Related Materials