EMAIL THIS PAGE TO A FRIEND
Organic letters
FeCl3 catalyzed Prins-type cyclization for the synthesis of highly substituted indenes: application to the total synthesis of (±)-jungianol and epi-jungianol.
PMID 23317410
Abstract
A novel approach was developed for the synthesis of highly substituted indene derivatives, using an FeCl(3) catalyzed Prins-type cyclization reaction which was further applied in the total synthesis of jungianol and epi-jungianol.