EMAIL THIS PAGE TO A FRIEND
Methods in cell biology
Immunohistochemical detection of neuropeptides/transmitters in mammalian brain.
PMID 23317901
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
441244
Paraformaldehyde, prilled, 95%
CH2O ]n