EMAIL THIS PAGE TO A FRIEND
Cell stem cell
Selective elimination of human pluripotent stem cells by an oleate synthesis inhibitor discovered in a high-throughput screen.
PMID 23318055
Abstract
The use of human pluripotent stem cells (hPSCs) in cell therapy is hindered by the tumorigenic risk from residual undifferentiated cells. Here we performed a high-throughput screen of over 52,000 small molecules and identified 15 pluripotent cell-specific inhibitors (PluriSIns), nine of which share a common structural moiety. The PluriSIns selectively eliminated hPSCs while sparing a large array of progenitor and differentiated cells. Cellular and molecular analyses demonstrated that the most selective compound, PluriSIn #1, induces ER stress, protein synthesis attenuation, and apoptosis in hPSCs. Close examination identified this molecule as an inhibitor of stearoyl-coA desaturase (SCD1), the key enzyme in oleic acid biosynthesis, revealing a unique role for lipid metabolism in hPSCs. PluriSIn #1 was also cytotoxic to mouse blastocysts, indicating that the dependence on oleate is inherent to the pluripotent state. Finally, application of PluriSIn #1 prevented teratoma formation from tumorigenic undifferentiated cells. These findings should increase the safety of hPSC-based treatments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML2356
A939572, ≥98% (HPLC)
C20H22ClN3O3
SML0682 PluriSIn #1, ≥95% (HPLC)
C12H11N3O