EMAIL THIS PAGE TO A FRIEND
The international journal of biochemistry & cell biology
Retinoic acid receptor beta mediates all-trans retinoic acid facilitation of mesenchymal stem cells neuronal differentiation.
PMID 23318218
Abstract
All-trans retinoic acid plays an important role in nervous system development. However, the effects of all-trans retinoic acid on the neuronal differentiation of mesenchymal stem cells and the mechanisms through which this differentiation takes place are still poorly understood. Here, we investigated the biological effects of all-trans retinoic acid on the neuronal differentiation of mesenchymal stem cells and the signaling pathways that mediated these effects. We found that the neuronal differentiation efficiency of mesenchymal stem cells following all-trans retinoic acid pre-induction was greater and the axonal length was longer than was observed with mesenchymal stem cells that were not pre-induced. mRNA and protein levels of the neural-markers Nestin, NSE, MAP-2, Tau and Tuj1 were stronger in neural-like cells derived from all-trans retinoic acid-pretreated mesenchymal stem cells than in those not pre-induction. Interestingly, the neuronal excitability of differentiated neural-like cells exhibited the same patterns between these two groups. Clear expression of retinoic acid receptor alpha and gamma in mesenchymal stem cells was observed, while retinoic acid receptor beta was barely detected. However, retinoic acid receptor beta expression in mesenchymal stem cells after neuronal induction increased dramatically, in contrast with retinoic acid receptor alpha and gamma expression, and retinoic acid receptor beta expression in mesenchymal stem cells receiving all-trans retinoic acid pre-induction was even stronger. Next, retinoic acid receptor alpha, beta and gamma were over-expressed by recombinant adenovirus infection prior to neuronal induction. Retinoic acid receptor alpha and gamma over-expression did not impact the neuronal differentiation of mesenchymal stem cells. However, retinoic acid receptor beta over-expression promoted neuronal differentiation to a similar level as observed following all-trans retinoic acid pre-induction. The neuronal differentiation promoting effects of all-trans retinoic acid on mesenchymal stem cells could be inhibited by siRNA silencing of retinoic acid receptor beta and by LE135, an inhibitor of retinoic acid receptor beta. Taken together, these results suggest that all-trans retinoic acid pre-induction facilitates the neuronal differentiation of mesenchymal stem cells. These facilitation effects are achieved by activating the retinoic acid receptor beta signaling pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0809 LE 135, ≥98% (HPLC)
C29H30N2O2