EMAIL THIS PAGE TO A FRIEND
FEBS letters
Molecular machines directly observed by high-speed atomic force microscopy.
PMID 23318713
Abstract
Molecular machines made of proteins are highly dynamic and carry out sophisticated biological functions. The direct and dynamic high-resolution visualization of molecular machines in action is considered to be the most straightforward approach to understanding how they function but this has long been infeasible until recently. High-speed atomic force microscopy has recently been realized, making such visualization possible. The captured images of myosin V, F1-ATPase, and bacteriorhodopsin have enabled their dynamic processes and structure dynamics to be revealed in great detail, giving unique and deep insights into their functional mechanisms.