EMAIL THIS PAGE TO A FRIEND
Annals of hematology
Hydroxyurea treatment in β-thalassemia patients: to respond or not to respond?
PMID 23318979
Abstract
Hydroxyurea (HU) is a drug that induces fetal hemoglobin production. As a result, HU is widely used to treat β-thalassemia (β-thal) patients. However, the response of these patients to HU varies. Some β-thal patients respond favorably to treatment while others do not respond at all. HU has a number of side-effects and therefore its targeted prescription is beneficial. Hence, identifying the genetic determinants which lead to the differential HU response is important. This review summarizes recent findings which have shed light on this topic. Special emphasis is given to the mechanisms and genetic loci which may govern these differences. These findings have helped identify several single nucleotide polymorphisms which associate with the response to HU in both β-thal and sickle cell disease patients.