EMAIL THIS PAGE TO A FRIEND
Macromolecular rapid communications
Combination of episulfide ring-opening polymerization with ATRP for the preparation of amphiphilic block copolymers.
PMID 23319175
Abstract
We report for the first time the combination of ATRP and ring-opening episulfide polymerization as a means to synthesize polysulfide-based low-dispersity amphiphilic block copolymers. The most significant finding is the possibility to perform ATRP under mild conditions using poly(propylene sulfide) macroinitiators, apparently without any significant copper sequestration by the polysulfides. Using glycerol monomethacrylate (GMMA) as a hydrophilic monomer, the polymers self-assembled in colloidal structures with a morphology depending on the PS/GMMA ratio, but also probably on GMMA degree of polymerization. We here also present a new AFM-based method to calculate the average number of amphiphilic macromolecules per micelle.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
82330
Propylene sulfide, purum, ≥96.0% (GC)
C3H6S
P53209
Propylene sulfide, ≥96%
C3H6S