EMAIL THIS PAGE TO A FRIEND
AAPS PharmSciTech
Development, characterization and permeability assessment based on caco-2 monolayers of self-microemulsifying floating tablets of tetrahydrocurcumin.
PMID 23319299
Abstract
Novel self-microemulsifying floating tablets were developed to enhance the dissolution and oral absorption of the poorly water-soluble tetrahydrocurcumin (THC). Their physicochemical properties and THC permeability across Caco-2 cell monolayers were assessed. The self-microemulsifying liquid containing THC was adsorbed onto colloidal silicon dioxide, mixed with HPMC, gas-generating agents (sodium bicarbonate and tartaric acid), lactose and silicified-microcrystalline cellulose and transformed into tablets by direct compression. The use of different types/concentrations of HPMC and sodium bicarbonate in tablet formulations had different effects on the floating characteristics and in vitro THC release. The optimum tablet formulation (F2) provided a short floating lag time (∼23 s) together with a prolonged buoyancy (>12 h). About 72% of THC was released in 12 h with an emulsion droplet size in aqueous media of 33.9±1.0 nm while that of a self-microemulsifying liquid was 29.9±0.3 nm. The tablet formulation was stable under intermediate and accelerated storage conditions for up to 6 months. The THC released from the self-microemulsifying liquid and tablet formulations provided an approximately three- to fivefold greater permeability across the Caco-2 cell monolayers than the unformulated THC and indicated an enhanced absorption of THC by the formulations. The self-microemulsifying floating tablet could provide a dosage form with the potential to improve the oral bioavailability of THC and other hydrophobic compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50202
Tetrahydrocurcumin, analytical standard
C21H24O6