EMAIL THIS PAGE TO A FRIEND
International journal of cancer
Human liver carboxylesterase 1 outperforms alpha-fetoprotein as biomarker to discriminate hepatocellular carcinoma from other liver diseases in Korean patients.
PMID 23319432
Abstract
Although alpha-fetoprotein (AFP) is currently the major serologic biomarker for hepatocellular carcinoma (HCC), it cannot efficiently distinguish this cancer from other forms of liver disease in early diagnosis due to its low sensitivity. The aim of this study is to compare sensitivity and specificity of human carboxylesterase 1 (hCE1) and AFP biomarker. Antibody-based assays for hCE1 and AFP were used to test both biomarkers with respect to diagnostic efficiency, Youden's index and the area under the curve (AUC) through receiver operating characteristic (ROC) analysis in plasma from 208 patients with HCC (n=57), liver cirrhosis (n=27), chronic hepatitis (n=37), cholangiocarcinoma (n=22), gastric cancer (n=31) and pancreatic cancer (n=34), along with 52 healthy donors (HDs). The levels of hCE1 were significantly higher in patients with HCC than HDs and the other diseases (p<0.005), further verified by AUC values and Youden's index. In the set of HCC versus liver cirrhosis the AUC values were 0.744 (AFP), 0.918 (hCE1) and 0.938 (combination of AFP and hCE1), respectively. These results indicate that hCE1 is not only a more potent and specific marker in distinguishing cancer from liver diseases, in particular cirrhosis, but the combination of hCE1 and AFP shows also synergistic potential for greater sensitivity and specificity in early diagnosis. Therefore the antibody-based hCE1 assay appears to have high diagnostic efficiency for discriminating HCC from other forms of liver disease. It is now feasible to further validate this novel plasma-based biomarker in the large cohort we assembled.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E0287 Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
E0162 Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
C4749 Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
46051 Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
79302 Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
96667 Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
51963 Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥0.8 U/mg
E3019 Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
E2884 Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
46058 Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
E0887 Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
79208 Esterase from Rhizopus oryzae, powder, light beige, ≥20 U/g
46071 Esterase from Saccharomyces cerevisiae, lyophilized, powder, beige, ~2 U/g
43388 Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
80712 Esterase Methylobacterium populi, recombinant, recombinant, expressed in E. coli, ≥0.5 U/mg
04492 Esterase Paenibacillus barcinonensis, recombinant, recombinant, expressed in E. coli, ≥10 U/mg
75742 Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg