EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Study on determination methods for abietic acid in fake Myrrha].
PMID 23320354
Abstract
To establish the qualitative and quantitative method for abietic acid in fake Myrrha. The abietic acid was identified by TLC, HPLC-PAD and HPLC-MS. Abietic acid was detected in 18 batches, which content was 1.16 to 73.99 (mg/g). The method can meet the requirements of identification and determination.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00010
Abietic acid, technical, ~75% (GC)
C20H30O2