EMAIL THIS PAGE TO A FRIEND
Current drug safety
Isolated unilateral tongue oedema: the adverse effect of Angiotensin converting enzyme inhibitors.
PMID 23320432
Abstract
Angiotensin converting enzyme inhibitors (ACEI) are widely used to treat benign hypertension. These drugs are generally well tolerated. Serious side effects such as angio-oedema are very rare. The authors report a 64-year-old Caucasian woman with a history of liver transplant on Mammalian Target Of Rapamycin (mTOR) inhibitor, who attended Emergency department with angio-oedema only on the left side of her tongue. Her airway was patent and she was haemodynamically stable. Trauma was denied. Her physician had 2 days earlier commenced her on Lisinopril for newly diagnosed benign hypertension. Intravenous steroids and anti-histamine were immediately administered. A good response of oedema subsidence was noted. In general, angio-oedema can present in a spectrum of severity. Precipitating factors are often difficult to pre-determine and avoid. Early recognition of risk factors for and diagnosis of angio-oedema can often be life-saving.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1143
Lisinopril, Pharmaceutical Secondary Standard; Certified Reference Material
C21H31O5N3 · 2H2O
L6292
Lisinopril, ≥98% (HPLC)
C21H31O5N3 · 2H2O