EMAIL THIS PAGE TO A FRIEND
Cytotherapy
Sildenafil improves long-term effect of endothelial progenitor cell-based treatment for monocrotaline-induced rat pulmonary arterial hypertension.
PMID 23321332
Abstract
We hypothesized that the long-term therapeutic effect of combined sildenafil and bone marrow-derived endothelial progenitor cells (BMDEPCs) on monocrotaline (MCT)-induced rat pulmonary arterial hypertension (PAH) is superior to either treatment alone. Male Sprague-Dawley rats (n = 40) were equally divided into normal controls, MCT (65 mg/kg, subcutaneously) only, MCT + sildenafil (25 mg/kg/day, orally), MCT + BMDEPCs (2.0 × 10(6) autologous cells, intravenously) and MCT + sildenafil+ BMDEPCs. BMDEPCs and sildenafil were given on day 21 after MCT administration. Animals were sacrificed by day 90 after MCT administration. The apoptotic (caspase 3, Bax) and inflammatory (tumor necrosis factor-α, matrix metalloproteinase-9) biomarkers in right ventricle and lung and pulmonary expressions of fibrotic biomarkers (transforming growth factor-β, p-Smad3) and connexin 43 protein were lower in monotherapy groups (i.e., MCT + sildenafil and MCT + BMDEPCs) and further decreased in normal controls and combined treatment groups (i.e., MCT + sildenafil + BMDEPCs) compared with untreated animals (i.e., MCT only) (all P < 0.01). Expressions of anti-fibrotic biomarkers (bone morphogenetic protein-2, p-Smad1/5) and numbers of alveolar sacs and arterioles in lung were higher in monotherapy groups and further increased in normal controls and combined treatment groups compared with untreated animals (all P < 0.005). In right ventricle, connexin 43 and α-myosin heavy chain (MHC) expressions were higher in the monotherapy groups and further elevated in normal controls and combined treatment groups compared with untreated animals, whereas β-MHC exhibited the opposite pattern (all P < 0.01). Right ventricular systolic pressure and weight were lower in the monotherapy animals and further reduced in normal controls and combined treatment groups compared with untreated animals (all P < 0.0001). Combined therapy with BMDEPCs and sildenafil was superior to either treatment alone in attenuating rodent MCT-induced PAH.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2401
Crotaline
C16H23NO6
37024
Monocrotaline, analytical standard
C16H23NO6