EMAIL THIS PAGE TO A FRIEND
European journal of clinical nutrition
The results of some studies involving animal models of obesity induced by monosodium glutamate are not conclusive.
PMID 23321571
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01495
L-Glutamic acid monosodium salt hydrate, analytical standard
C5H8NNaO4 · xH2O