EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Sciences
Expression of partitioning defective 3 (Par-3) for predicting extrahepatic metastasis and survival with hepatocellular carcinoma.
PMID 23322019