EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Template-free bottom-up synthesis of yolk-shell vanadium oxide as high performance cathode for lithium ion batteries.
PMID 23322132
Abstract
A template-free strategy is exploited to bottom-up synthesize yolk-shell vanadium oxide through a two-step spontaneous assembly of hydrolytically formed subunits in a one-pot process. The unique structured vanadium pentoxide exhibits excellent cathode performance for lithium ion batteries.