EMAIL THIS PAGE TO A FRIEND
Muscle & nerve
Muscle cramps and weakness after teriparatide therapy: a new drug-induced myopathy?
PMID 23322619
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3796 Parathyroid Hormone Fragment 1-34 human, ≥95% (HPLC), powder
C181H291N55O51S2