EMAIL THIS PAGE TO A FRIEND
Scientific reports
Novel selective and irreversible mosquito acetylcholinesterase inhibitors for controlling malaria and other mosquito-borne diseases.
PMID 23323211
Abstract
We reported previously that insect acetylcholinesterases (AChEs) could be selectively and irreversibly inhibited by methanethiosulfonates presumably through conjugation to an insect-specific cysteine in these enzymes. However, no direct proof for the conjugation has been published to date, and doubts remain about whether such cysteine-targeting inhibitors have desirable kinetic properties for insecticide use. Here we report mass spectrometric proof of the conjugation and new chemicals that irreversibly inhibited African malaria mosquito AChE with bimolecular inhibition rate constants (k(inact)/K(I)) of 3,604-458,597 M(-1)sec(-1) but spared human AChE. In comparison, the insecticide paraoxon irreversibly inhibited mosquito and human AChEs with k(inact)/K(I) values of 1,915 and 1,507 M(-1)sec(-1), respectively, under the same assay conditions. These results further support our hypothesis that the insect-specific AChE cysteine is a unique and unexplored target to develop new insecticides with reduced insecticide resistance and low toxicity to mammals, fish, and birds for the control of mosquito-borne diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36186
Paraoxon-ethyl, PESTANAL®, analytical standard
C10H14NO6P