EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[Effect of 3-oxypyridine and succinic acid derivatives on affective status in recrudescence of inflammatory diseases of uterus and its appendages].
PMID 23323329
Abstract
Short-term, prospective placebo-controlled simple blind randomized study of the effects of 3-oxypyridine and succinic acid derivatives (emoxipin, reamberin, mexidol) on the affective status of females with recrudescence of the inflammatory diseases of uterus and its appendages (IDUA) in comparison to changes of systemic inflammatory response (SIR) markers level in the blood has been conducted. It is established that the inclusion of emoxipin, reamberin and mexidol in complex treatment of IDUA recrudescence reduce depression, anxiety and SIR laboratory signs. Mexidol being both 3-oxypyridine and succinic acid derivative showed the best influence on the dynamics of affective disorders and SIR changes.