EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Slow magnetization relaxation in unprecedented Mn(III)4Dy(III)3 and Mn(III)4Dy(III)5 clusters from the use of N-salicylidene-o-aminophenol.
PMID 23323592
Abstract
The first use of N-salicylidene-o-aminophenol in 3d/4f chemistry has led to Mn(III)(4)Dy(III)(5) and Mn(III)(4)Dy(III)(3) clusters with unprecedented metal topologies and stoichiometries; both compounds exhibit out-of-phase signals indicative of the slow magnetization relaxation of a single-molecule magnet.