EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
High nuclearity complexes of lanthanide involving tetrathiafulvalene ligands: structural, magnetic, and photophysical properties.
PMID 23323869
Abstract
The reaction between the tetrakis(2-pyridyl-N-oxidemethylthio)tetrathiafulvalene ligand (L) and Ln(hfac)(3)·2H(2)O precursors (where hfac(-) = 1,1,1,5,5,5-hexafluoroacetylacetonate anion and Ln = Tb(III) (1), Dy(III) (2), Er(III) (3), and Yb(III) (4) and (4b)) leads to the formation of five tetranuclear complexes of formula [Ln(4)(hfac)(12)(L)(2)](n)·xCHCl(3)·yC(6)H(14) (n = 1, x = 2, y = 0 for (1), (2), and (4), n = 1, x = 4 for (3), and n = 2, x = 2.5, y = 1 for (4b)). Their X-ray structures reveal that the surrounding of each Ln(III) center is filled by two N-oxide groups coming from two different ligands L. These tetranuclear complexes have the highest nuclearity which is reported until now for coordination compounds of lanthanide involving TTF-based ligands. Direct current (dc) measurements highlight the paramagnetic behavior of the compounds with a significant crystal field effect. The temperature dependences of static magnetic measurements for 4 have been fitted. The ground state corresponds to M(J) = ±5/2 while the first excited state (M(J) = ±3/2) was localized at +214 cm(-1) which was well correlated with the luminescence transition. UV-visible absorption properties have been experimentally measured and rationalized by time-dependent density functional theory (TD-DFT) calculations. Upon irradiation at 77 K and room temperature, in the range 24390-20835 cm(-1), both compounds 3 and 4 display a metal-centered luminescence attributed to (4)I(13/2) → (4)I(15/2) (6660 cm(-1)) and (2)F(5/2) → (2)F(7/2) (signal centered around the value of 9966 cm(-1)) transitions, respectively. The observed six transitions could be attributed to the M(J) state splitting due to the existence of two Yb1 and Yb2 ions with slightly different polyhedra in 4.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
183180
Tetrathiafulvalene, 97%
C6H4S4