EMAIL THIS PAGE TO A FRIEND
Organic letters
Quantum chemical calculations of 1J(CC) coupling constants for the stereochemical determination of organic compounds.
PMID 23324108
Abstract
Quantum chemical calculations of one-bond carbon-carbon coupling constants are demonstrated as potential probes for the configurational assignment of organic molecules. The stereochemical analysis of strychnine and its possible stereoisomers is presented as proof of concept.