EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Adduct formation identification between phenyl glycidyl ether and 2'-deoxyadenosine and thymidine by chromatography, mass spectrometry and nuclear magnetic resonance spectroscopy.
PMID 2332460
Abstract
Thymidine and 2'-deoxyadenosine were reacted with phenyl glycidyl ether in order to study the formation of the corresponding 2'-deoxynucleoside adducts. Separation methods were elaborated using either reversed-phase high-performance liquid chromatography with photodiode-array detection, or centrifugal circular thin-layer chromatography. The adducts were isolated on a preparative scale and were fully characterized by UV spectroscopy, desorption chemical ionization and fast atom bombardment mass spectrometry and 270- and 360-MHz 1H NMR spectrometry. For thymidine the main adduct was characterized as N-3-(2-hydroxy-3-phenoxypropyl)thymidine. With 2'-deoxyadenosine, predominantly N-1-(2-hydroxy-3-phenoxypropyl)-2'-deoxyadenosine was formed. With longer reaction times, the formation of a minor amount of dialkylated 2'-deoxyadenosine was observed. These nucleoside adducts will be used as marker compounds for studies of DNA adduct formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
248487
1,2-Epoxy-3-phenoxypropane, 99%
C9H10O2
578576
1,2-Epoxy-3-phenoxypropane, technical grade
C9H10O2