EMAIL THIS PAGE TO A FRIEND
JAMA surgery
Is the elimination of postoperative antimicrobial prophylaxis in thoracic surgery ready for prime time?: Comment on "Postoperative antibacterial prophylaxis for the prevention of infectious complications associated with tube thoracostomy in patients undergoing elective general thoracic surgery".
PMID 23324804