EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Improvement of the enzyme performance of trypsin via adsorption in mesoporous silica SBA-15: hydrolysis of BAPNA.
PMID 23325102
Abstract
The enzymatic performance of trypsin in hydrolysis of N-α-benzoyl-DL-arginine-4-nitroanilide (BAPNA) was improved by adsorption on Santa Barbara Amorphous (SBA)-15 mesoporous silica. The optimal immobilization conditions were screened and the properties of immobilized enzyme have also been studied. Under the optimal conditions, the immobilized trypsin displays maximum specific activity (49.8 μmol/min/g). The results also indicate that the immobilized trypsin exhibits better storage stability.