EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Mesoporous silica nanoparticle facilitated drug release through cascade photosensitizer activation and cleavage of singlet oxygen sensitive linker.
PMID 23325385
Abstract
A photoresponsive drug releasing system based on mesoporous silica nanoparticles (MSN) was developed. A model drug was decorated on the surfaces via singlet oxygen sensitive linkers and a singlet oxygen-generating photosensitizer was loaded in the mesopores of the MSN. An on-demand drug releasing system was successfully developed by irradiation with long-wavelength light.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
341169
Zinc phthalocyanine, Dye content 97 %
C32H16N8Zn